Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APG3
Entrez Gene ID
825466
Gene Name
Peptide chain release factor 1
Gene Aliases
ALBINO AND PALE GREEN, AT3G62910, Arabidopsis thaliana chloroplast RF1, AtcpRF1, chloroplast ribosome release factor 1, cpRF1
Chromosome Location
Chr.3: 23257404 - 23260480 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116156.5 NP_191850.2 3 - 4 388 110
GenBank mRNA AB109893.1 - 3 - 4 294 110
AY090253.1 - 3 - 4 335 110
AY125535.1 - 3 - 4 294 110

View Full Product Details