Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPX5
Entrez Gene ID
825483
Gene Name
glutathione peroxidase 5
Gene Aliases
AT3G63080, ATGPX5, MEE42, glutathione peroxidase 5, maternal effect embryo arrest 42
Chromosome Location
Chr.3: 23309737 - 23311397 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116173.4 NP_191867.1 3 - 4 622 65
GenBank mRNA AY085116.1 - 3 - 4 245 65
BT003821.1 - 3 - 4 254 65
BT005137.1 - 3 - 4 198 65

View Full Product Details