Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TKL
Entrez Gene ID
825493
Gene Name
TIC-like protein
Gene Aliases
AT3G63180, ATTKL, TIC-like, TICKLE
Chromosome Location
Chr.3: 23336092 - 23340340 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084865.2 NP_001078334.1 2 - 3 529 65
NM_116183.4 NP_191877.2 2 - 3 552 65

View Full Product Details