Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RRF
Entrez Gene ID
825494
Gene Name
ribosome recycling factor, chloroplast precursor
Gene Aliases
''ribosome recycling factor, AT3G63190, Arabidopsis thaliana chloroplast ribosome recycling factor, AtcpRRF, HFP108, HIGH CHLOROPHYLL FLUORESCENCE AND PALE GREEN MUTANT 108, chloroplast precursor, chloroplast precursor'', chloroplast ribosome recycling factor, cpRRF, ribosome recycling factor
Chromosome Location
Chr.3: 23342550 - 23344801 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116184.4 NP_567141.1 10 - 11 972 139
GenBank mRNA AF372883.1 - 10 - 11 860 139
AY056778.1 - 10 - 11 940 139
AY092982.1 - 10 - 11 878 139

View Full Product Details