Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NEK4
Entrez Gene ID
825503
Gene Name
NIMA-related kinase 4
Gene Aliases
AT3G63280, ATNEK4, MAA21.6, NIMA-RELATED KINASE4, NIMA-related kinase 4
Chromosome Location
Chr.3: 23377976 - 23381590 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125427.1 NP_001118899.1 7 - 8 523 85
NM_001340197.1 NP_001326587.1 8 - 9 562 85
NM_001340198.1 NP_001326588.1 7 - 8 562 85
NM_116193.5 NP_191887.2 8 - 9 603 85
GenBank mRNA AY080682.1 - 8 - 9 597 85
AY117234.1 - 6 - 7 386 85

View Full Product Details