Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CKX6
- Entrez Gene ID
-
825519
- Gene Name
- cytokinin oxidase/dehydrogenase 6
- Gene Aliases
-
AT3G63440, ATCKX6, ATCKX7, CYTOKININ OXIDASE 6, cytokinin oxidase/dehydrogenase 6
- Chromosome Location
-
Chr.3: 23424157 - 23426536 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_116209.3 |
NP_191903.3 |
3 - 4 |
1150 |
86 |
View Full Product Details