Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CCD1
- Entrez Gene ID
-
825527
- Gene Name
- carotenoid cleavage dioxygenase 1
- Gene Aliases
-
AT3G63520, ATCCD1, ATNCED1, CAROTENOID CLEAVAGE DIOXYGENASE 1, NCED1, carotenoid cleavage dioxygenase 1
- Chromosome Location
-
Chr.3: 23452808 - 23456078 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_116217.3 |
NP_191911.1 |
2 - 3 |
418 |
89 |
View Full Product Details