Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UNE10
Entrez Gene ID
828175
Gene Name
basic helix-loop-helix (bHLH) DNA-binding superfamily protein
Gene Aliases
AT4G00050, F6N15.11, F6N15_11, unfertilized embryo sac 10
Chromosome Location
Chr.4: 17639 - 20183 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340226.1 NP_001329601.1 3 - 4 886 155
NM_001340227.1 NP_001329602.1 4 - 5 811 155
NM_116222.4 NP_191916.3 4 - 5 956 155
GenBank mRNA AK117229.1 - 4 - 5 803 155
BT025971.1 - 4 - 5 732 155

View Full Product Details