Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MEE44
- Entrez Gene ID
-
828214
- Gene Name
- Nucleotidyltransferase family protein
- Gene Aliases
-
AT4G00060, F6N15.10, F6N15_10, maternal effect embryo arrest 44
- Chromosome Location
-
Chr.4: 21351 - 29064 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_116223.4 |
NP_191917.3 |
16 - 17 |
4681 |
89 |
View Full Product Details