Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MEE45
- Entrez Gene ID
-
828225
- Gene Name
- Transcriptional factor B3 family protein
- Gene Aliases
-
AT4G00260, A_IG005I10.15, A_IG005I10_15, F5I10.15, F5I10_15, maternal effect embryo arrest 45
- Chromosome Location
-
Chr.4: 114971 - 117051 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_116247.2 |
NP_567162.2 |
6 - 7 |
1282 |
150 |
View Full Product Details