Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANTR2
Entrez Gene ID
827908
Gene Name
Major facilitator superfamily protein
Gene Aliases
AT4G00370, A_IG005I10.NN, A_IG005I10_NN, F5I10.7, F5I10_7, PHT4;4, anion transporter 2
Chromosome Location
Chr.4: 162897 - 166357 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116261.4 NP_567175.2 3 - 4 695 88
GenBank mRNA AK118390.1 - 3 - 4 570 88
BT009663.1 - 3 - 4 449 88

View Full Product Details