The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_116268.4 | NP_567178.1 | 1 - 2 | 478 | 71 |
NM_202760.2 | NP_974489.1 | 1 - 2 | 425 | 71 | |
GenBank mRNA | AY099825.1 | - | 1 - 2 | 403 | 71 |
AY120785.1 | - | 1 - 2 | 407 | 71 | |
BT000330.1 | - | 1 - 2 | 329 | 71 | |
BT006313.1 | - | 1 - 2 | 329 | 71 | |
D26609.1 | - | 1 - 2 | 462 | 71 |