Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATMYC1
Entrez Gene ID
827965
Gene Name
basic helix-loop-helix (bHLH) DNA-binding superfamily protein
Gene Aliases
AT4G00480, F6N23.22, F6N23_22, myc1
Chromosome Location
Chr.4: 216812 - 219926 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160722.2 NP_001154194.1 1 - 2 244 147
NM_001340255.1 NP_001329692.1 1 - 2 280 147
NM_116272.4 NP_191957.2 1 - 2 389 147
GenBank mRNA AB493664.1 - 1 - 2 147 147
AF251697.1 - 1 - 2 147 147

View Full Product Details