Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ECA2
Entrez Gene ID
827984
Gene Name
ER-type Ca2+-ATPase 2
Gene Aliases
ARABIDOPSIS THALIANA ER-TYPE CA2+-ATPASE 2, AT4G00900, ATECA2, A_TM018A10.4, A_TM018A10_4, ER-type Ca2+-ATPase 2, T18A10.20, T18A10_20
Chromosome Location
Chr.4: 382663 - 386850 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340276.1 NP_001329712.1 4 - 5 2608 94
NM_116317.4 NP_191999.1 5 - 6 2783 94
GenBank mRNA AJ132387.1 - 4 - 5 2417 94

View Full Product Details