Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIT1
Entrez Gene ID
828146
Gene Name
RING/FYVE/PHD zinc finger superfamily protein
Gene Aliases
AT4G02075, pitchoun 1
Chromosome Location
Chr.4: 913334 - 916509 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340359.1 NP_001329350.1 1 - 2 228 71
NM_001340360.1 NP_001329351.1 1 - 2 228 71
NM_116439.3 NP_567222.1 1 - 2 228 71
GenBank mRNA AF130849.1 - 1 - 2 195 71
DQ086841.1 - 1 - 2 133 71

View Full Product Details