Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GH9B13
Entrez Gene ID
828080
Gene Name
glycosyl hydrolase 9B13
Gene Aliases
AT4G02290, AtGH9B13, T2H3.5, T2H3_5, glycosyl hydrolase 9B13
Chromosome Location
Chr.4: 1002338 - 1005305 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340370.1 NP_001328883.1 4 - 5 1347 111
NM_116462.4 NP_192138.1 4 - 5 1120 111
GenBank mRNA AY079162.1 - 4 - 5 1018 111
AY101518.1 - 4 - 5 940 111

View Full Product Details