Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CUL1
Entrez Gene ID
825648
Gene Name
cullin 1
Gene Aliases
AT4G02570, ATCUL1, AUXIN RESISTANT 6, AXR6, T10P11.26, T10P11_26, cullin 1
Chromosome Location
Chr.4: 1128515 - 1133774 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036498.3 NP_001031575.1 11 - 12 1220 127
NM_001036499.1 NP_001031576.1 11 - 12 1156 127
NM_001203732.1 NP_001190661.1 11 - 12 1218 127
NM_116491.3 NP_567243.1 10 - 11 1314 127
GenBank mRNA AJ318017.1 - 9 - 10 1015 127
AK317123.1 - 11 - 12 1157 127
AY046030.1 - 10 - 11 1237 127
AY133878.1 - 9 - 10 981 127

View Full Product Details