Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- GA1
- Entrez Gene ID
-
828182
- Gene Name
- Terpenoid cyclases/Protein prenyltransferases superfamily protein
- Gene Aliases
-
ABC33, ARABIDOPSIS THALIANA ENT-COPALYL DIPHOSPHATE SYNTHETASE 1, AT4G02780, ATCPS1, CPP synthase, CPS, CPS1, ENT-COPALYL DIPHOSPHATE SYNTHETASE, ENT-COPALYL DIPHOSPHATE SYNTHETASE 1, GA REQUIRING 1, T5J8.9, T5J8_9
- Chromosome Location
-
Chr.4: 1237671 - 1244822 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_116512.2 |
NP_192187.1 |
1 - 2 |
151 |
111 |
GenBank mRNA |
U11034.1 |
- |
1 - 2 |
142 |
111 |
View Full Product Details