Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABP1
Entrez Gene ID
828120
Gene Name
endoplasmic reticulum auxin binding protein 1
Gene Aliases
ABP, AT-ERABP1, AT4G02980, T4I9.14, T4I9_14, endoplasmic reticulum auxin binding protein 1
Chromosome Location
Chr.4: 1319603 - 1321608 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116532.3 NP_192207.1 4 - 5 645 71
GenBank mRNA AF389278.1 - 4 - 5 495 71
AY093754.1 - 4 - 5 486 71
S40550.1 - 4 - 5 495 71

View Full Product Details