Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DFL2
Entrez Gene ID
827938
Gene Name
Auxin-responsive GH3 family protein
Gene Aliases
AT4G03400, DWARF IN LIGHT 2, F9H3.1, GH3-10
Chromosome Location
Chr.4: 1497536 - 1501599 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340446.1 NP_001319858.1 4 - 5 2010 84
NM_116578.3 NP_192249.1 2 - 3 778 84
GenBank mRNA AY059941.1 - 2 - 3 542 84
BT000061.1 - 2 - 3 407 84

View Full Product Details