Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GSL05
- Entrez Gene ID
-
825650
- Gene Name
- glucan synthase-like 5
- Gene Aliases
-
AT4G03550, ATGSL05, ATGSL5, EED3, GLUCAN SYNTHASE-LIKE 5, GSL5, PMR4, POWDERY MILDEW RESISTANT 4, T5L23.4, enhancer of edr1 3, glucan synthase-like 5
- Chromosome Location
-
Chr.4: 1573266 - 1579820 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_116593.4 |
NP_192264.1 |
1 - 2 |
2111 |
70 |
GenBank mRNA |
KJ138766.1 |
- |
1 - 2 |
1864 |
70 |
View Full Product Details