Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FIB
Entrez Gene ID
825714
Gene Name
fibrillin
Gene Aliases
AT4G04020, FIB1a, PGL35, T24H24.16, T24H24_16, fibrillin, fibrillin 1a, plastoglobulin 35
Chromosome Location
Chr.4: 1931883 - 1933950 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116640.5 NP_192311.1 1 - 2 726 67
GenBank mRNA AY085330.1 - 1 - 2 459 67
AY120766.2 - 1 - 2 459 67
BT000148.1 - 1 - 2 448 67

View Full Product Details