Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MEE51
- Entrez Gene ID
-
825716
- Gene Name
- Phosphofructokinase family protein
- Gene Aliases
-
AT4G04040, T24H24.15, T24H24_15, maternal effect embryo arrest 51
- Chromosome Location
-
Chr.4: 1939113 - 1943024 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_116642.4 |
NP_192313.3 |
7 - 8 |
952 |
92 |
View Full Product Details