Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALN
Entrez Gene ID
825836
Gene Name
allantoinase
Gene Aliases
AT4G04955, ATALN, allantoinase
Chromosome Location
Chr.4: 2522034 - 2525352 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116734.3 NP_567276.1 1 - 2 187 110
GenBank mRNA AY045901.1 - 1 - 2 115 110
AY113948.1 - 1 - 2 94 110

View Full Product Details