Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GSL1
- Entrez Gene ID
-
825838
- Gene Name
- glucan synthase-like 1
- Gene Aliases
-
AT4G04970, ATGSL01, ATGSL1, GLUCAN SYNTHASE LIKE 1, GLUCAN SYNTHASE LIKE-1, GSL01, T32N4.8, T32N4_8, glucan synthase-like 1
- Chromosome Location
-
Chr.4: 2536925 - 2543273 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_116736.2 |
NP_567278.1 |
3 - 4 |
5659 |
104 |
View Full Product Details