Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDB2
Entrez Gene ID
825844
Gene Name
NAD(P)H dehydrogenase B2
Gene Aliases
AT4G05020, C17L7.11, NAD(P)H dehydrogenase B2
Chromosome Location
Chr.4: 2572427 - 2576599 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203749.1 NP_001190678.1 6 - 7 1269 88
NM_116741.4 NP_567283.1 6 - 7 1463 88
GenBank mRNA AY039856.1 - 6 - 7 1195 88
BT002241.1 - 6 - 7 1138 88

View Full Product Details