Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATK5
Entrez Gene ID
825867
Gene Name
kinesin 5
Gene Aliases
AT4G05190, C17L7.110, C17L7_110, kinesin 5
Chromosome Location
Chr.4: 2675064 - 2680214 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340542.1 NP_001329202.1 7 - 8 1099 89
NM_116758.4 NP_192428.2 8 - 9 1396 89

View Full Product Details