Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EHD2
Entrez Gene ID
825902
Gene Name
EPS15 homology domain 2
Gene Aliases
AT4G05520, ATEHD2, EPS15 homology domain 2, T1J24.10, T1J24_10
Chromosome Location
Chr.4: 2804403 - 2808015 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116790.3 NP_567299.2 7 - 8 582 130
NM_202788.1 NP_974517.1 7 - 8 531 130
GenBank mRNA BT033036.1 - 7 - 8 463 130

View Full Product Details