Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IBR1
Entrez Gene ID
825905
Gene Name
indole-3-butyric acid response 1
Gene Aliases
AT4G05530, INCREASED BOTRYTIS RESISTANCE 1, SDRA, SHORT-CHAIN DEHYDROGENASE/REDUCTASE A, T1J24.9, T1J24_9, indole-3-butyric acid response 1
Chromosome Location
Chr.4: 2816317 - 2818518 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116791.3 NP_567300.1 1 - 2 276 59
GenBank mRNA AY035106.1 - 1 - 2 181 59
AY087807.1 - 1 - 2 198 59
AY113900.1 - 1 - 2 131 59

View Full Product Details