Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ORP1C
Entrez Gene ID
826368
Gene Name
OSBP(oxysterol binding protein)-related protein 1C
Gene Aliases
AT4G08180, OSBP(oxysterol binding protein)-related protein 1C, T12G13.20, T12G13_20
Chromosome Location
Chr.4: 5169459 - 5174107 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340578.1 NP_001319881.1 4 - 5 1725 87
NM_116887.3 NP_192558.2 4 - 5 1700 87
NM_179012.2 NP_849343.2 4 - 5 1697 87
NM_202789.1 NP_974518.1 4 - 5 1697 87
GenBank mRNA AY139750.1 - 4 - 5 1698 87

View Full Product Details