Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GTA2
- Entrez Gene ID
-
826391
- Gene Name
- global transcription factor group A2
- Gene Aliases
-
AT4G08350, GTA02, SPT5-2, T28D5.40, T28D5_40, global transcription factor group A2
- Chromosome Location
-
Chr.4: 5286158 - 5292270 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_116904.6 |
NP_192575.3 |
14 - 15 |
2452 |
109 |
View Full Product Details