Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRZ3
Entrez Gene ID
841647
Gene Name
tRNAse Z3
Gene Aliases
AT1G52160, F9I5.1, F9I5_1, tRNAse Z3
Chromosome Location
Chr.1: 19420715 - 19424474 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104097.6 NP_175628.2 5 - 6 604 83
GenBank mRNA AY070061.1 - 5 - 6 569 83
AY117303.1 - 5 - 6 549 83

View Full Product Details