Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MSL4
Entrez Gene ID
841782
Gene Name
mechanosensitive channel of small conductance-like 4
Gene Aliases
AT1G53470, T3F20.21, T3F20_21, mechanosensitive channel of small conductance-like 4
Chromosome Location
Chr.1: 19958380 - 19961827 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104225.2 NP_175752.1 1 - 2 1929 88

View Full Product Details