Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAP3KA
Entrez Gene ID
841792
Gene Name
mitogen-activated protein kinase kinase kinase 3
Gene Aliases
AT1G53570, F22G10.18, MAP KINASE KINASE KINASE 3, MAPKKK3, MEK KINASE, mitogen-activated protein kinase kinase kinase 3
Chromosome Location
Chr.1: 19987117 - 19991214 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036104.2 NP_001031181.1 6 - 7 1288 97
NM_001198282.1 NP_001185211.1 6 - 7 1288 97
NM_001333597.1 NP_001322087.1 6 - 7 1362 97
NM_001333598.1 NP_001322088.1 6 - 7 1313 97
NM_104235.4 NP_564635.1 6 - 7 1317 97
NM_179472.3 NP_849803.1 6 - 7 1362 97
GenBank mRNA AY140005.1 - 6 - 7 1213 97
BT002598.1 - 6 - 7 1088 97

View Full Product Details