Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPT1A
Entrez Gene ID
841812
Gene Name
regulatory particle triple-A 1A
Gene Aliases
26S PROTEASOME ATPASE SUBUNIT, AT1G53750, T18A20.1, T18A20_1, regulatory particle triple-A 1A
Chromosome Location
Chr.1: 20065394 - 20068500 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104252.3 NP_175778.1 4 - 5 509 97
GenBank mRNA AY062860.1 - 4 - 5 473 97
BT000069.1 - 4 - 5 333 97

View Full Product Details