Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAE1
Entrez Gene ID
841822
Gene Name
20S proteasome alpha subunit E1
Gene Aliases
20S PROTEASOME SUBUNIT PAE1, 20S proteasome alpha subunit E1, ARABIDOPSIS 20S PROTEASOME ALPHA SUBUNIT E1, AT1G53850, ATPAE1, T18A20.8, T18A20_8
Chromosome Location
Chr.1: 20103756 - 20106077 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084248.2 NP_001077717.1 7 - 8 702 79
NM_104262.3 NP_175788.1 8 - 9 743 79
GenBank mRNA AF043524.1 - 8 - 9 639 79
AY072371.1 - 8 - 9 659 79
AY086045.1 - 8 - 9 661 79
AY114616.1 - 6 - 7 546 79

View Full Product Details