Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXO70D2
Entrez Gene ID
841847
Gene Name
exocyst subunit exo70 family protein D2
Gene Aliases
AT1G54090, ATEXO70D2, F15I1.17, F15I1_17, exocyst subunit exo70 family protein D2
Chromosome Location
Chr.1: 20189472 - 20191900 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104286.3 NP_175811.1 1 - 1 2377 111
GenBank mRNA AY093196.1 - 1 - 1 2286 111

View Full Product Details