Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATG18H
Entrez Gene ID
841912
Gene Name
autophagy 18h-like protein
Gene Aliases
AT1G54710, ATATG18H, T22H22.14, T22H22_14, homolog of yeast autophagy 18 (ATG18) H
Chromosome Location
Chr.1: 20416770 - 20421156 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333664.1 NP_001323099.1 5 - 6 2091 113
NM_104346.3 NP_564664.1 6 - 7 2672 113
GenBank mRNA AY142626.1 - 6 - 7 2249 113

View Full Product Details