Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NP2
Entrez Gene ID
841937
Gene Name
NPK1-related protein kinase 2
Gene Aliases
ANP2, AT1G54960, F14C21.49, F14C21_49, MAPKKK2, MITOGEN-ACTIVATED PROTEIN KINASE KINASE KINASES 2, NPK1-related protein kinase 2
Chromosome Location
Chr.1: 20499776 - 20504076 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333684.1 NP_001319236.1 13 - 14 1461 123
NM_104370.5 NP_175894.5 13 - 14 1437 123

View Full Product Details