Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IDD7
Entrez Gene ID
841954
Gene Name
indeterminate(ID)-domain 7
Gene Aliases
AT1G55110, AtIDD7, T7N22.5, T7N22_5, indeterminate(ID)-domain 7
Chromosome Location
Chr.1: 20560258 - 20563269 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333688.1 NP_001322628.1 1 - 2 861 119
NM_001333689.1 NP_001322629.1 2 - 3 702 119
NM_104384.4 NP_175907.2 2 - 3 405 119
GenBank mRNA AK227194.1 - 2 - 3 285 119
AY150421.1 - 1 - 2 217 119

View Full Product Details