Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FRUCT5
Entrez Gene ID
841955
Gene Name
beta-fructofuranosidase 5
Gene Aliases
6-fructan exohydrolase, AT1G55120, ATFRUCT5, AtcwINV3, BETA-FRUCTOFURANOSIDASE, T7N22.6, T7N22_6, beta-fructofuranosidase 5
Chromosome Location
Chr.1: 20566030 - 20569322 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001124022.2 NP_001117494.1 3 - 4 1304 80
NM_001198301.1 NP_001185230.1 3 - 4 1183 80
NM_001333690.1 NP_001320753.1 3 - 4 1480 80
NM_104385.4 NP_564676.1 4 - 5 1324 80

View Full Product Details