Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDF5
Entrez Gene ID
841984
Gene Name
NDH-dependent cyclic electron flow 5
Gene Aliases
AT1G55370, NDH-dependent cyclic electron flow 5, T18I3.2
Chromosome Location
Chr.1: 20673768 - 20676449 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104413.4 NP_564687.1 1 - 2 1324 65
NM_179479.3 NP_849810.1 1 - 2 1324 65
GenBank mRNA AK227298.1 - 1 - 2 313 65

View Full Product Details