Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAX5
Entrez Gene ID
842022
Gene Name
cation exchanger 5
Gene Aliases
AT1G55730, ATCAX5, F20N2.30, F20N2_30, cation exchanger 5
Chromosome Location
Chr.1: 20831031 - 20834628 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036119.2 NP_001031196.1 6 - 7 953 82
NM_104449.3 NP_175969.2 6 - 7 860 82
GenBank mRNA AK317482.1 - 6 - 7 835 82
AY136416.1 - 6 - 7 842 82
BT006591.1 - 5 - 6 588 82

View Full Product Details