Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DI19
Entrez Gene ID
842081
Gene Name
drought-induced 19
Gene Aliases
AT1G56280, ATDI19, F14G9.11, F14G9_11, drought-induced 19
Chromosome Location
Chr.1: 21072855 - 21074877 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333775.1 NP_001319253.1 4 - 5 889 77
NM_104507.4 NP_564715.4 4 - 5 643 77

View Full Product Details