Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PBG1
Entrez Gene ID
842098
Gene Name
20S proteasome beta subunit G1
Gene Aliases
20S proteasome beta subunit G1, AT1G56450, F13N6.3, F13N6_3, PROTEASOME BETA SUBUNIT PBG1
Chromosome Location
Chr.1: 21141633 - 21144458 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104523.4 NP_176040.1 4 - 5 616 91
GenBank mRNA AF378880.1 - 4 - 5 355 91
AK221762.1 - 4 - 5 366 91
AY045596.1 - 4 - 5 332 91
AY052739.1 - 3 - 4 279 91

View Full Product Details