The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_104527.4 | NP_176043.1 | 1 - 2 | 604 | 105 |
GenBank mRNA | AK226771.1 | - | 1 - 2 | 586 | 105 |
BT014857.1 | - | 1 - 2 | 465 | 105 |