The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_104590.4 | NP_176105.1 | 6 - 7 | 969 | 89 |
GenBank mRNA | AB025249.1 | - | 6 - 7 | 724 | 89 |
AY093018.1 | - | 6 - 7 | 743 | 89 | |
AY128941.1 | - | 6 - 7 | 719 | 89 |