Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RLP9
Entrez Gene ID
842186
Gene Name
receptor like protein 9
Gene Aliases
AT1G58190, AtRLP9, T18I24.10, T18I24_10, receptor like protein 9
Chromosome Location
Chr.1: 21540584 - 21548119 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198331.1 NP_001185260.1 2 - 3 597 116
NM_104600.3 NP_176115.2 2 - 3 645 116

View Full Product Details