Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSTU15
Entrez Gene ID
842257
Gene Name
glutathione S-transferase TAU 15
Gene Aliases
AT1G59670, ATGSTU15, T30E16.25, T30E16_25, glutathione S-transferase TAU 15
Chromosome Location
Chr.1: 21930010 - 21931366 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104660.2 NP_176176.1 1 - 2 353 61
GenBank mRNA DQ446376.1 - 1 - 2 328 61

View Full Product Details