Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALA3
Entrez Gene ID
842275
Gene Name
aminophospholipid ATPase 3
Gene Aliases
AT1G59820, F23H11.14, F23H11_14, aminophospholipid ATPase 3
Chromosome Location
Chr.1: 22011305 - 22020386 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104675.3 NP_176191.1 18 - 19 2078 76
GenBank mRNA AY091777.1 - 18 - 19 2055 76

View Full Product Details